Gene Transcripts

Gene Symbol
Eif3j
Entrez Gene ID
691947
Gene Name
eukaryotic translation initiation factor 3, subunit J
Gene Aliases
Eif3s1
Chromosome Location
Chr. 3: 113976687 - 113998925 on Build Rnor_6.0
UniGene ID
Rn.162681
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077670.1 NP_001071138.1 5 488
GenBank mRNA BC127509.1 - 5 488
FQ226401.1 - 5 437

View Full Product Details