Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007204.4 | NP_009135.4 | 11 | 1742 | |
GenBank mRNA | AF106019.1 | - | 11 | 1395 | |
AF155803.1 | - | 11 | 1390 | ||
AF171063.1 | - | 11 | 1678 | ||
AK001506.1 | - | 11 | 1404 | ||
AK301697.1 | - | 10 | 1243 | ||
AK314312.1 | - | 11 | 1387 | ||
AL133598.1 | - | 3 | 2566 | ||
BC011556.1 | - | 11 | 1395 | 4651697 | |
BC031062.1 | - | 11 | 1404 | 5273709 | |
BC034953.2 | - | 11 | 1713 | 4824324 |