Gene Transcripts

Gene Symbol
PHB2
Entrez Gene ID
11331
Gene Name
prohibitin 2
Gene Aliases
BAP, BCAP37, Bap37, PNAS-141, REA, hBAP, p22
Chromosome Location
Chr. 12: 6965352 - 6970823 on Build GRCh38
UniGene ID
Hs.504620
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144831.1 NP_001138303.1 8 1022
NM_001267700.1 NP_001254629.1 7 908
GenBank mRNA AF126021.1 - 8 997
AF150962.1 - 8 935
AF178980.1 - 8 911
AF279895.1 - 3 123
AK298217.1 - 7 828
AK301316.1 - 7 1098
BC013228.1 - 8 905
BC014766.1 - 8 892 3452360
BC110322.1 - 8 1022 5540527
U72511.1 - 6 550

View Full Product Details