Gene Transcripts

Gene Symbol
UTP11
Entrez Gene ID
51118
Gene Name
UTP11, small subunit processome component homolog (S. cerevisiae)
Gene Aliases
CGI-94, CGI94, UTP11L
Chromosome Location
Chr. 1: 38012712 - 38024825 on Build GRCh38
UniGene ID
Hs.472038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016037.3 NP_057121.2 7 757
XM_011541555.2 XP_011539857.1 6 691
GenBank mRNA AF067802.1 - 7 720
AF151852.1 - 7 735
AK023587.1 - 5 1626
AK291900.1 - 7 687
AK296018.1 - 6 559
AK308377.1 - 6 692
BC005182.1 - 7 702 3995984
CX163866.1 - 2 123

View Full Product Details