Gene Transcripts

Gene Symbol
PRICKLE4
Entrez Gene ID
29964
Gene Name
prickle planar cell polarity protein 4
Gene Aliases
C6orf49, OBTP, OEBT, TOMM6
Chromosome Location
Chr. 6: 41780762 - 41787372 on Build GRCh38
UniGene ID
Hs.715653
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013397.5 NP_037529.3 8 1051
GenBank mRNA BC110458.1 - 6 1498
BC110459.1 - 6 1503 40024767

Gene Symbol
TOMM6
Entrez Gene ID
100188893
Gene Name
translocase of outer mitochondrial membrane 6
Gene Aliases
OBTP, TOM6
Chromosome Location
Chr. 6: 41787443 - 41789896 on Build GRCh38
UniGene ID
Hs.731633
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ420506.1 - 1 44

View Full Product Details