Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000026.3 | NP_000017.1 | 6 | 735 | |
NM_001123378.2 | NP_001116850.1 | 6 | 735 | ||
NM_001317923.1 | NP_001304852.1 | 6 | 680 | ||
NR_134256.1 | 6 | 735 | |||
XM_011529977.2 | XP_011528279.1 | 6 | 735 | ||
XM_011529979.1 | XP_011528281.1 | 6 | 735 | ||
XM_011529980.2 | XP_011528282.1 | 6 | 735 | ||
XM_017028636.1 | XP_016884125.1 | 5 | 672 | ||
XM_017028637.1 | XP_016884126.1 | 5 | 655 | ||
XM_017028638.1 | XP_016884127.1 | 4 | 592 | ||
XM_017028639.1 | XP_016884128.1 | 4 | 592 | ||
XM_017028640.1 | XP_016884129.1 | 2 | 90 | ||
GenBank mRNA | AF067853.1 | - | 6 | 731 | |
AF067854.1 | - | 6 | 731 | ||
AK225042.1 | - | 6 | 703 | ||
AK291091.1 | - | 6 | 706 | ||
AK298187.1 | - | 6 | 749 | ||
AK300705.1 | - | 4 | 512 | ||
AK308662.1 | - | 2 | 90 | ||
BC000253.1 | - | 6 | 705 | 3352079 | |
BC018881.2 | - | 6 | 705 | ||
CR456368.1 | - | 6 | 709 | ||
KJ534763.1 | - | 6 | 676 | ||
KJ535007.1 | - | 6 | 676 | ||
KJ535020.1 | - | 6 | 676 | ||
KJ535041.1 | - | 3 | 347 | ||
KJ535056.1 | - | 6 | 621 | ||
S60710.1 | - | 6 | 678 | ||
X65867.1 | - | 6 | 678 |