Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081259.1 | NP_001074728.1 | 7, 8 | 1529 | |
GenBank mRNA | AK036315.1 | - | 6, 7 | 1126 | |
AK165872.1 | - | 7, 8 | 1529 | ||
BC024751.1 | - | 2, 3 | 507 | ||
BC157992.1 | - | 7, 8 | 1497 | 9007305 |