Gene Transcripts

Gene Symbol
ZC3H7B
Entrez Gene ID
23264
Gene Name
zinc finger CCCH-type containing 7B
Gene Aliases
RoXaN
Chromosome Location
Chr. 22: 41301503 - 41360147 on Build GRCh38
UniGene ID
Hs.592188
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017590.5 NP_060060.3 5 650
XM_011530016.2 XP_011528318.1 5 631
GenBank mRNA AB028954.1 - 2 263
AF188530.1 - 5 478
AK023849.1 - 5 438
AK225891.1 - 5 623
AK315009.1 - 5 592
BC098378.1 - 5 588
BC152558.1 - 5 628 8860405

View Full Product Details