Gene Transcripts

Gene Symbol
Spry1
Entrez Gene ID
294981
Gene Name
sprouty homolog 1, antagonist of FGF signaling (Drosophila)
Gene Aliases
Chromosome Location
Chr. 2: 124400689 - 124405331 on Build Rnor_6.0
UniGene ID
Rn.22787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106427.1 NP_001099897.1 2 267
GenBank mRNA BC169059.1 - 3 566

View Full Product Details