Gene Transcripts

Gene Symbol
ACSL6
Entrez Gene ID
23305
Gene Name
acyl-CoA synthetase long-chain family member 6
Gene Aliases
ACS2, FACL6, LACS 6, LACS2, LACS5
Chromosome Location
Chr. 5: 131949973 - 132012068 on Build GRCh38
UniGene ID
Hs.14945
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009185.2 NP_001009185.1 5 621
NM_001205247.1 NP_001192176.1 5 516
NM_001205248.1 NP_001192177.1 5 608
NM_001205250.1 NP_001192179.1 5 618
NM_001205251.1 NP_001192180.1 5 503
NM_015256.3 NP_056071.2 5 621
GenBank mRNA AB020644.1 - 5 581
AF102845.1 - 4 398
AF129166.1 - 5 551
AK294291.1 - 5 491
AK314452.1 - 5 523
BC026161.1 - 5 517 4830357
BC047453.1 - 5 425 5270879
BI464352.1 - 5 604
BI545119.1 - 5 622
DQ083031.1 - 4 437

View Full Product Details