Gene Transcripts

Gene Symbol
Vmac
Entrez Gene ID
363327
Gene Name
vimentin-type intermediate filament associated coiled-coil protein
Gene Aliases
Chromosome Location
Chr. 9: 10338245 - 10341903 on Build Rnor_6.0
UniGene ID
Rn.214255
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001720.1 NP_001001720.1 1, 2 190
XM_008766765.1 XP_008764987.1 2, 3 290
GenBank mRNA AY521229.1 - 1, 2 190

View Full Product Details