Gene Transcripts

Gene Symbol
GCA
Entrez Gene ID
25801
Gene Name
grancalcin
Gene Aliases
GCL
Chromosome Location
Chr. 2: 162318768 - 162371605 on Build GRCh38
UniGene ID
Hs.377894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012198.3 NP_036330.1 2 336
XM_005246446.2 XP_005246503.1 2 277
XM_006712398.3 XP_006712461.1 2 509
XM_006712400.3 XP_006712463.1 3 587
XM_006712401.3 XP_006712464.1 2 588
XM_011510926.2 XP_011509228.1 2 288
XM_011510927.2 XP_011509229.1 2 283
XM_017003764.1 XP_016859253.1 3 600
XM_017003765.1 XP_016859254.1 3 686
XM_017003766.1 XP_016859255.1 3 483
XM_017003767.1 XP_016859256.1 2 359
XM_017003768.1 XP_016859257.1 2 354
XM_017003769.1 XP_016859258.1 2 509
GenBank mRNA AB209768.1 - 2 225
AK295413.1 - 2 269
AK312349.1 - 2 285
BC005214.1 - 2 259 3996889
M81637.1 - 2 279

View Full Product Details