Gene Transcripts

Gene Symbol
SMC1B
Entrez Gene ID
27127
Gene Name
structural maintenance of chromosomes 1B
Gene Aliases
SMC1BETA, SMC1L2
Chromosome Location
Chr. 22: 45344063 - 45413671 on Build GRCh38
UniGene ID
Hs.334176
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291501.1 NP_001278430.1 15 2414
NM_148674.4 NP_683515.4 15 2414
XM_011530144.2 XP_011528446.1 15 2275
XM_011530145.2 XP_011528447.1 15 2466
GenBank mRNA AJ504806.2 - 15 2414
AK125736.1 - 2 173
BC126208.1 - 15 2394 8991924

View Full Product Details