Gene Transcripts

Gene Symbol
SREBF2
Entrez Gene ID
6721
Gene Name
sterol regulatory element binding transcription factor 2
Gene Aliases
SREBP-2, SREBP2, bHLHd2
Chromosome Location
Chr. 22: 41833079 - 41907308 on Build GRCh38
UniGene ID
Hs.443258
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004599.3 NP_004590.2 15 2885
NR_103834.1 16 2985
XM_006724310.2 XP_006724373.1 15 3129
XM_011530347.1 XP_011528649.1 13 2339
XM_017028921.1 XP_016884410.1 15 2714
GenBank mRNA AK299914.1 - 15 2714
AL834221.1 - 7 1244
BC051385.1 - 16 2891 5497929
BC051799.1 - 15 2851
BC056158.1 - 15 2831 6169568
CT841522.1 - 15 2775
U02031.1 - 15 2810

View Full Product Details