Gene Transcripts

Gene Symbol
MRPS18C
Entrez Gene ID
51023
Gene Name
mitochondrial ribosomal protein S18C
Gene Aliases
CGI-134, MRP-S18-1, MRP-S18-c, MRPS18-1, S18mt-c, mrps18-c
Chromosome Location
Chr. 4: 83455932 - 83461723 on Build GRCh38
UniGene ID
Hs.334772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297769.1 NP_001284698.1 3 364
NM_016067.3 NP_057151.1 3 364
GenBank mRNA AF151892.1 - 3 277
AK002008.1 - 2 1617
AK026620.1 - 3 331
BC005186.1 - 3 255 3839381
BE874586.1 - 3 206

View Full Product Details