Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017425.3 | NP_059121.1 | 3 | 339 | |
XM_011542870.2 | XP_011541172.1 | 3 | 529 | ||
GenBank mRNA | AF334735.1 | - | 3 | 338 | |
AK311806.1 | - | 3 | 282 | ||
BC019224.1 | - | 3 | 271 | ||
BC032457.2 | - | 2 | 231 | 5171849 | |
DA511916.1 | - | 3 | 388 | ||
KU257683.1 | - | 2 | 203 |