Gene Transcripts

Gene Symbol
PRR13
Entrez Gene ID
54458
Gene Name
proline rich 13
Gene Aliases
TXR1
Chromosome Location
Chr. 12: 53441649 - 53446643 on Build GRCh38
UniGene ID
Hs.744894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005354.2 NP_001005354.1 4 664
NM_018457.3 NP_060927.1 4 814
GenBank mRNA AF217517.1 - 1 727
AK074398.1 - 1 710
BC001464.2 - 4 800
BC004512.2 - 1 407
BC014257.2 - 1 714 4765821
BC016064.1 - 1 711 4852862
BC021019.1 - 1 682
BC066943.1 - 1 713 4817871
BC071979.1 - 1 645
BX640962.1 - 2 552

View Full Product Details