Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142549.1 | NP_001136021.1 | 12 | 798 | |
NM_001166533.1 | NP_001160005.1 | 12 | 840 | ||
NM_001166534.1 | NP_001160006.1 | 6 | 399 | ||
NM_024415.2 | NP_077726.1 | 13 | 900 | ||
XM_011543495.2 | XP_011541797.1 | 12 | 845 | ||
XM_011543497.2 | XP_011541799.1 | 11 | 785 | ||
GenBank mRNA | AF262962.1 | - | 12 | 818 | |
AK093439.1 | - | 6 | 373 | ||
AK292417.1 | - | 13 | 875 | ||
AK308300.1 | - | 12 | 817 | ||
AL137462.1 | - | 9 | 536 | ||
AY004154.1 | - | 12 | 814 | ||
BC047455.1 | - | 12 | 736 | 5298807 | |
BC088362.1 | - | 13 | 859 | 30386875 | |
DB022413.1 | - | 7 | 445 | ||
DB051674.1 | - | 7 | 448 |