Gene Transcripts
- Gene Symbol
- C2orf42
- Entrez Gene ID
- 54980
- Gene Name
- chromosome 2 open reading frame 42
- Gene Aliases
- Chromosome Location
- Chr. 2: 70149885 - 70191071 on Build GRCh38
- UniGene ID
- Hs.720049
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017880.1 |
NP_060350.1
|
8, 9 |
1687 |
|
XM_005264389.2 |
XP_005264446.1
|
8, 9 |
1880 |
|
XM_005264390.2 |
XP_005264447.1
|
8, 9 |
1749 |
|
XM_005264391.2 |
XP_005264448.1
|
8, 9 |
1831 |
|
XM_011532937.2 |
XP_011531239.1
|
7, 8 |
1625 |
|
XM_017004398.1 |
XP_016859887.1
|
8, 9 |
2168 |
|
XM_017004399.1 |
XP_016859888.1
|
8, 9 |
1876 |
|
XM_017004400.1 |
XP_016859889.1
|
8, 9 |
1760 |
|
GenBank mRNA |
AK000565.1 |
- |
8, 9 |
1687 |
|
AK026303.1 |
- |
6, 7 |
972 |
|
AL136834.1 |
- |
7, 8 |
1465 |
|
BC002825.2 |
- |
8, 9 |
1676 |
3635986 |
BC005079.1 |
- |
8, 9 |
1676 |
3615724 |
View Full Product Details