Gene Transcripts
- Gene Symbol
- SMU1
- Entrez Gene ID
- 55234
- Gene Name
- DNA replication regulator and spliceosomal factor
- Gene Aliases
- BWD, SMU-1, fSAP57
- Chromosome Location
- Chr. 9: 33041852 - 33076716 on Build GRCh38
- UniGene ID
- Hs.655351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018225.2 |
NP_060695.2
|
11, 12 |
1552 |
|
XM_005251503.4 |
XP_005251560.1
|
10, 11 |
1388 |
|
GenBank mRNA |
AK001667.1 |
- |
11, 12 |
1482 |
|
AK022032.1 |
- |
3, 4 |
356 |
|
AK225559.1 |
- |
11, 12 |
1507 |
|
AK225587.1 |
- |
11, 12 |
1501 |
|
AK304767.1 |
- |
9, 10 |
1139 |
|
BC002876.2 |
- |
11, 12 |
1498 |
3940669 |
BC110654.1 |
- |
11, 12 |
1592 |
6168993 |
EF011613.1 |
- |
11, 12 |
1498 |
|
View Full Product Details