Gene Transcripts

Gene Symbol
ITLN1
Entrez Gene ID
55600
Gene Name
intelectin 1
Gene Aliases
HL-1, HL1, INTL, ITLN, LFR, hIntL, omentin
Chromosome Location
Chr. 1: 160876540 - 160885170 on Build GRCh38
UniGene ID
Hs.50813
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017625.2 NP_060095.2 2, 3 180
GenBank mRNA AB036706.1 - 2, 3 92
AF271386.1 - 2, 3 103
AK000029.1 - 2, 3 129
AK222533.1 - 2, 3 167
AY065972.1 - 2, 3 170
AY157361.1 - 2, 3 182
AY157362.1 - 2, 3 174
AY358359.1 - 2, 3 166
AY549722.1 - 2, 3 269
AY619691.1 - 2, 3 90
AY619692.1 - 2, 3 147
BC020664.1 - 2, 3 180 4693329
CR457224.1 - 1, 2 64

View Full Product Details