Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291277.1 | NP_001278206.1 | 2 | 209 | |
NM_031274.4 | NP_112564.1 | 2 | 235 | ||
GenBank mRNA | AF285597.1 | - | 2 | 216 | |
BC042547.1 | - | 2 | 190 | 5164310 | |
BC108734.1 | - | 2 | 208 | 5744723 | |
HY018872.1 | - | 2 | 209 |