Gene Transcripts

Gene Symbol
TTYH1
Entrez Gene ID
57348
Gene Name
tweety family member 1
Gene Aliases
Chromosome Location
Chr. 19: 54415431 - 54436719 on Build GRCh38
UniGene ID
Hs.268728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005367.2 NP_001005367.1 10 1192
NM_001201461.1 NP_001188390.1 10 1192
NM_020659.3 NP_065710.1 10 1192
XM_017027025.1 XP_016882514.1 10 1036
XM_017027026.1 XP_016882515.1 10 1036
GenBank mRNA AB162930.1 - 10 1075
AB188496.1 - 10 1192
AF177909.1 - 10 1106
AK126690.1 - 9 1103
AK289468.1 - 10 1155
AK293468.1 - 11 1223
BC011347.1 - 10 1097 4215136
BC019358.1 - 7 525 3960435

View Full Product Details