Gene Transcripts

Gene Symbol
NFE4
Entrez Gene ID
58160
Gene Name
nuclear factor, erythroid 4
Gene Aliases
NF-E4
Chromosome Location
Chr. 7: 102973437 - 102978667 on Build GRCh38
UniGene ID
Hs.621233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085386.1 NP_001078855.1 1 162
GenBank mRNA AY258907.1 - 1 162
BC036938.1 - 1 88

View Full Product Details