Gene Transcripts

Gene Symbol
RRAGC
Entrez Gene ID
64121
Gene Name
Ras related GTP binding C
Gene Aliases
GTR2, RAGC, TIB929
Chromosome Location
Chr. 1: 38838197 - 38859823 on Build GRCh38
UniGene ID
Hs.532461
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271851.1 NP_001258780.1 7 1148
NM_022157.3 NP_071440.1 7 1250
GenBank mRNA AF272035.1 - 7 1078
AF323609.1 - 7 1073
AK023373.1 - 3 278
AK092808.1 - 2 2161
AK298573.1 - 6 773
AK298788.1 - 7 1044
AK315791.1 - 7 1174
BC016668.1 - 7 1095 4342448
CX761032.1 - 2 160

View Full Product Details