Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204414.1 | NP_001191343.1 | 3 | 480 | |
NM_001204415.1 | NP_001191344.1 | 2 | 88 | ||
NM_022164.2 | NP_071447.1 | 3 | 480 | ||
XM_005271106.2 | XP_005271163.1 | 3 | 480 | ||
XM_005271107.2 | XP_005271164.1 | 2 | 125 | ||
GenBank mRNA | AB050716.1 | - | 3 | 430 | |
AF205436.1 | - | 3 | 406 | ||
AF236150.1 | - | 3 | 408 | ||
AK074124.1 | - | 2 | 469 | ||
AK075398.1 | - | 3 | 453 | ||
AK292770.1 | - | 3 | 454 | ||
AK296322.1 | - | 3 | 450 | ||
AK298382.1 | - | 3 | 450 | ||
AK301070.1 | - | 3 | 490 | ||
AY358421.1 | - | 3 | 453 | ||
BC009048.1 | - | 11 | |||
BC064633.1 | - | 3 | 438 | 5750149 | |
BM923842.1 | - | 2 | 88 | ||
DA656570.1 | - | 3 | 482 |