Gene Transcripts

Gene Symbol
PDGFD
Entrez Gene ID
80310
Gene Name
platelet derived growth factor D
Gene Aliases
IEGF, MSTP036, SCDGF-B, SCDGFB
Chromosome Location
Chr. 11: 103907186 - 104164299 on Build GRCh38
UniGene ID
Hs.352298
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025208.4 NP_079484.1 6 1205
NM_033135.3 NP_149126.1 6 1187
GenBank mRNA AB033832.1 - 6 989
AF113216.1 - 6 939
AF335584.1 - 6 1014
AF336376.1 - 6 1008
AK292801.1 - 6 1013
AY027517.1 - 6 925
AY027518.1 - 6 907
AY359116.1 - 6 846
BC030645.2 - 6 1267 4824526

View Full Product Details