Gene Transcripts
- Gene Symbol
- OR2M3
- Entrez Gene ID
- 127062
- Gene Name
- olfactory receptor family 2 subfamily M member 3
- Gene Aliases
- OR1-54, OR2M3P, OR2M6, OST003
- Chromosome Location
- Chr. 1: 248203068 - 248204006 on Build GRCh38
- UniGene ID
- Hs.553581
- Species
- Homo sapiens
- Gene Symbol
- OR2M1P
- Entrez Gene ID
- 388762
- Gene Name
- olfactory receptor family 2 subfamily M member 1 pseudogene
- Gene Aliases
- JCG10, OR2M1, OST037
- Chromosome Location
- Chr. 1: 248122136 - 248122780 on Build GRCh38
- UniGene ID
- Hs.528509
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002141.1 |
|
1 |
473 |
|
- Gene Symbol
- OR2M2
- Entrez Gene ID
- 391194
- Gene Name
- olfactory receptor family 2 subfamily M member 2
- Gene Aliases
- OR2M2Q, OST423
- Chromosome Location
- Chr. 1: 248179986 - 248181029 on Build GRCh38
- UniGene ID
- Hs.534728
- Species
- Homo sapiens
- Gene Symbol
- OR2M5
- Entrez Gene ID
- 127059
- Gene Name
- olfactory receptor family 2 subfamily M member 5
- Gene Aliases
- OR2M5P
- Chromosome Location
- Chr. 1: 248145148 - 248146086 on Build GRCh38
- UniGene ID
- Hs.553580
- Species
- Homo sapiens
- Gene Symbol
- OR2M7
- Entrez Gene ID
- 391196
- Gene Name
- olfactory receptor family 2 subfamily M member 7
- Gene Aliases
- OR1-58
- Chromosome Location
- Chr. 1: 248323630 - 248324568 on Build GRCh38
- UniGene ID
- Hs.530570
- Species
- Homo sapiens
View Full Product Details