Gene Transcripts

Gene Symbol
SAMD13
Entrez Gene ID
148418
Gene Name
sterile alpha motif domain containing 13
Gene Aliases
HSD-41, HSD-42
Chromosome Location
Chr. 1: 84298366 - 84350798 on Build GRCh38
UniGene ID
Hs.591445
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010971.2 NP_001010971.1 3 372
NM_001134663.1 NP_001128135.1 3 366
NM_001134664.1 NP_001128136.1 3 210
XM_017000377.1 XP_016855866.1 3 1040
GenBank mRNA AK056937.1 - 3 372
BC037973.1 - 3 210
DB459148.2 - 3 367

View Full Product Details