Gene Transcripts

Gene Symbol
WDR49
Entrez Gene ID
151790
Gene Name
WD repeat domain 49
Gene Aliases
Chromosome Location
Chr. 3: 167478684 - 167653501 on Build GRCh38
UniGene ID
Hs.213762
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178824.3 NP_849146.1 10 1597
GenBank mRNA AK090939.1 - 8 1002
AK294387.1 - 10 1518
BC035512.2 - 10 1597 5199615

View Full Product Details