Gene Transcripts
- Gene Symbol
- LCE1E
- Entrez Gene ID
- 353135
- Gene Name
- late cornified envelope 1E
- Gene Aliases
- LEP5
- Chromosome Location
- Chr. 1: 152786277 - 152788426 on Build GRCh38
- UniGene ID
- Hs.250236
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178353.1 |
NP_848130.1
|
1, 2 |
42 |
|
GenBank mRNA |
BC038391.1 |
- |
1, 2 |
42 |
|
View Full Product Details