Gene Transcripts

Gene Symbol
LCLAT1
Entrez Gene ID
253558
Gene Name
lysocardiolipin acyltransferase 1
Gene Aliases
1AGPAT8, AGPAT8, ALCAT1, HSRG1849, LYCAT, UNQ1849
Chromosome Location
Chr. 2: 30447226 - 30644225 on Build GRCh38
UniGene ID
Hs.468048
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002257.2 NP_001002257.1 3 357
NM_001304445.1 NP_001291374.1 5 762
NM_001304446.1 NP_001291375.1 2 209
NM_182551.4 NP_872357.2 4 549
XM_005264244.1 XP_005264301.1 4 511
XM_005264245.3 XP_005264302.1 3 363
XM_011532741.2 XP_011531043.1 4 500
XM_011532742.2 XP_011531044.1 4 327
XM_011532743.2 XP_011531045.1 3 314
XM_011532744.2 XP_011531046.1 3 352
XM_011532745.1 XP_011531047.1 4 511
XM_017003746.1 XP_016859235.1 4 394
XM_017003747.1 XP_016859236.1 4 511
GenBank mRNA AK095284.1 - 4 514
AK296965.1 - 3 342
AK297262.1 - 2 189
AK302562.1 - 5 715
AY358702.1 - 4 518
BC146817.1 - 3 347 8860419
BP872689.1 - 3 357
BX647953.1 - 3 313

View Full Product Details