Gene Transcripts

Gene Symbol
SLC24A5
Entrez Gene ID
283652
Gene Name
solute carrier family 24 member 5
Gene Aliases
JSX, NCKX5, OCA6, SHEP4
Chromosome Location
Chr. 15: 48120972 - 48142392 on Build GRCh38
UniGene ID
Hs.710240
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205850.2 NP_995322.1 3 443
XM_017022079.1 XP_016877568.1 3 1451
XM_017022080.1 XP_016877569.1 3 1671
GenBank mRNA AF348468.1 - 3 443
BC113628.1 - 3 398 8322680
BC113630.1 - 3 398 8322682
BC143950.1 - 3 398 9052468
DQ665306.1 - 3 391
DQ665307.1 - 2 211

View Full Product Details