Gene Transcripts

Gene Symbol
TAS2R60
Entrez Gene ID
338398
Gene Name
taste 2 receptor member 60
Gene Aliases
T2R56, T2R60
Chromosome Location
Chr. 7: 143443453 - 143444409 on Build GRCh38
UniGene ID
Hs.688222
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177437.1 NP_803186.1 1 531
GenBank mRNA BC100938.2 - 1 531
BC100939.2 - 1 531 40004057
BC100940.1 - 1 531 40004058

View Full Product Details