Gene Transcripts

Gene Symbol
SBK1
Entrez Gene ID
388228
Gene Name
SH3 domain binding kinase 1
Gene Aliases
SBK
Chromosome Location
Chr. 16: 28258687 - 28323849 on Build GRCh38
UniGene ID
Hs.97837
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024401.2 NP_001019572.1 2, 3 1013
XM_005255315.3 XP_005255372.1 2, 3 1047
GenBank mRNA AY874862.2 - 2, 3 1013

View Full Product Details