Gene Transcripts

Gene Symbol
OR6X1
Entrez Gene ID
390260
Gene Name
olfactory receptor family 6 subfamily X member 1
Gene Aliases
OR11-270
Chromosome Location
Chr. 11: 123753580 - 123754518 on Build GRCh38
UniGene ID
Hs.553759
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005188.1 NP_001005188.1 1 330
GenBank mRNA BC136855.1 - 1 357
BC136856.1 - 1 357 9020847

View Full Product Details