Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013254.3 | NP_037386.1 | 17, 18 | 2016 | |
XM_005268809.1 | XP_005268866.1 | 17, 18 | 2094 | ||
XM_005268810.1 | XP_005268867.1 | 17, 18 | 2076 | ||
GenBank mRNA | AF174536.1 | - | 16, 17 | 1857 | |
AF191838.1 | - | 17, 18 | 1919 | ||
AK002192.1 | - | 9, 10 | 842 | ||
AK291039.1 | - | 17, 18 | 1961 | ||
AK303683.1 | - | 12, 13 | 1233 | ||
BC034950.2 | - | 17, 18 | 1956 | 4824988 |