Gene Transcripts
- Gene Symbol
- NDUFB3
- Entrez Gene ID
- 4709
- Gene Name
- NADH:ubiquinone oxidoreductase subunit B3
- Gene Aliases
- B12, CI-B12
- Chromosome Location
- Chr. 2: 201071739 - 201085750 on Build GRCh38
- UniGene ID
- Hs.109760
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001257102.1 |
NP_001244031.1
|
3, 4 |
494 |
|
NM_002491.2 |
NP_002482.1
|
2, 3 |
471 |
|
XM_011511230.2 |
XP_011509532.1
|
3, 4 |
508 |
|
XM_017004186.1 |
XP_016859675.1
|
2, 3 |
716 |
|
GenBank mRNA |
AF035839.1 |
- |
2, 3 |
296 |
|
AF047183.1 |
- |
2, 3 |
400 |
|
BC018183.1 |
- |
2, 3 |
419 |
3881592 |
BM848639.1 |
- |
2, 3 |
471 |
|
BU188560.1 |
- |
2, 3 |
198 |
|
CR456924.1 |
- |
1, 2 |
148 |
|
View Full Product Details