Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109878.1 | NP_001103348.1 | 9 | 1462 | |
NM_001109879.1 | NP_001103349.1 | 9 | 1458 | ||
NM_001303475.1 | NP_001290404.1 | 7 | 1216 | ||
NM_016954.2 | NP_058650.1 | 8 | 1355 | ||
XM_005262136.2 | XP_005262193.1 | 9 | 1490 | ||
XM_011530972.1 | XP_011529274.1 | 8 | 2095 | ||
GenBank mRNA | AF251684.1 | - | 7 | 1207 | |
AK097514.1 | - | 9 | 1458 | ||
AY035371.1 | - | 8 | 1334 | ||
BC014194.2 | - | 8 | 1355 | 3952809 |