Gene Transcripts

Gene Symbol
UNC13C
Entrez Gene ID
440279
Gene Name
unc-13 homolog C
Gene Aliases
Chromosome Location
Chr. 15: 53837573 - 54628608 on Build GRCh38
UniGene ID
Hs.657273
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080534.1 NP_001074003.1 22, 23 5388
XM_005254394.4 XP_005254451.1 23, 24 6339
XM_017022220.1 XP_016877709.1 27, 28 6259
XM_017022221.1 XP_016877710.1 26, 27 6139
XM_017022222.1 XP_016877711.1 25, 26 6037
XM_017022223.1 XP_016877712.1 23, 24 11565
XM_017022224.1 XP_016877713.1 22, 23 6265
XM_017022225.1 XP_016877714.1 20, 21 3690
GenBank mRNA AK091491.1 - 19, 20 2364
AK293996.1 - 12, 13 1305
AL834407.1 - 6, 7 663

View Full Product Details