Gene Transcripts

Gene Symbol
UBE4B
Entrez Gene ID
10277
Gene Name
ubiquitination factor E4B
Gene Aliases
E4, HDNB1, UBOX3, UFD2, UFD2A
Chromosome Location
Chr. 1: 10032983 - 10181239 on Build GRCh38
UniGene ID
Hs.593974
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105562.2 NP_001099032.1 5, 6 1273
NM_006048.4 NP_006039.2 5, 6 1273
XM_005263422.1 XP_005263479.1 5, 6 1097
XM_011540488.2 XP_011538790.1 4, 5 396
XM_011540489.2 XP_011538791.1 5, 6 1097
XM_011540490.1 XP_011538792.1 5, 6 1097
XM_017000035.1 XP_016855524.1 2, 3 183
XM_017000036.1 XP_016855525.1 6, 7 937
GenBank mRNA AB014584.3 - 5, 6 1164
AB028839.1 - 5, 6 1281
AF043117.1 - 5, 6 670
AF331520.1 - 5, 6 632
AK292444.1 - 5, 6 755
AK304484.1 - 3, 4 260
BC093696.1 - 5, 6 663 7939541
BC143458.1 - 5, 6 663
BC143460.1 - 5, 6 663
JF289274.1 - 5, 6 585

View Full Product Details