Gene Transcripts

Gene Symbol
ZAR1L
Entrez Gene ID
646799
Gene Name
zygote arrest 1-like
Gene Aliases
Z3CXXC7, ZAR2
Chromosome Location
Chr. 13: 32303771 - 32311954 on Build GRCh38
UniGene ID
Hs.569254
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136571.1 NP_001130043.1 3 830
GenBank mRNA BC146956.1 - 3 828
BC146965.1 - 3 830 9056893

View Full Product Details