Gene Transcripts

Gene Symbol
Rhoa
Entrez Gene ID
11848
Gene Name
ras homolog gene family, member A
Gene Aliases
Arha, Arha1, Arha2
Chromosome Location
Chr. 9: 108306205 - 108337939 on Build GRCm38
UniGene ID
Mm.318359
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016802.4 NP_058082.2 5 800
GenBank mRNA AF014371.1 - 4 508
AF178958.1 - 4 395
AF178959.1 - 4 395
AF178960.1 - 4 395
AF178961.1 - 4 395
AK077606.1 - 5 608
AK083624.1 - 5 607
AK150294.1 - 5 645
AK150475.1 - 5 651
AK152629.1 - 5 587
AK165512.1 - 5 583
AK172051.1 - 5 608
BC068115.1 - 5 800 6809149
BC096423.1 - 5 586 3590185
JN971019.1 - 3 374

View Full Product Details