Gene Transcripts

Gene Symbol
HOXC10
Entrez Gene ID
3226
Gene Name
homeobox C10
Gene Aliases
HOX3I
Chromosome Location
Chr. 12: 53985162 - 53990279 on Build GRCh38
UniGene ID
Hs.44276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017409.3 NP_059105.2 2 1074
GenBank mRNA AF255675.1 - 2 976
AK313759.1 - 2 1090
BC001293.1 - 2 1047 3458115
BT009921.1 - 2 976

View Full Product Details