Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002160.3 | NP_002151.2 | 4 | 2369 | |
XM_005251972.3 | XP_005252029.1 | 4 | 2319 | ||
XM_005251973.3 | XP_005252030.1 | 4 | 2319 | ||
XM_005251974.3 | XP_005252031.1 | 4 | 2319 | ||
XM_005251975.3 | XP_005252032.1 | 4 | 2319 | ||
XM_006717096.3 | XP_006717159.1 | 4 | 2319 | ||
XM_006717097.3 | XP_006717160.1 | 4 | 2319 | ||
XM_006717098.3 | XP_006717161.1 | 4 | 2319 | ||
XM_006717101.3 | XP_006717164.1 | 4 | 2319 | ||
XM_011518625.2 | XP_011516927.1 | 4 | 2319 | ||
XM_011518626.2 | XP_011516928.1 | 4 | 2319 | ||
XM_011518628.2 | XP_011516930.1 | 4 | 2319 | ||
XM_011518629.2 | XP_011516931.1 | 4 | 2319 | ||
XM_017014678.1 | XP_016870167.1 | 4 | 2319 | ||
XM_017014679.1 | XP_016870168.1 | 4 | 2319 | ||
XM_017014680.1 | XP_016870169.1 | 4 | 2319 | ||
XM_017014681.1 | XP_016870170.1 | 4 | 2319 | ||
GenBank mRNA | AB210038.1 | - | 4 | 2245 | |
AK304015.1 | - | 4 | 2265 | ||
BC151843.1 | - | 4 | 2237 | 8860362 | |
GU473235.1 | - | 3 | 1957 | ||
M24630.1 | - | 2 | 667 | ||
M55618.1 | - | 3 | 2092 | ||
X56160.1 | - | 3 | 2008 | ||
X78565.1 | - | 4 | 2270 |