Gene Transcripts

Gene Symbol
Pafah1b1
Entrez Gene ID
18472
Gene Name
platelet-activating factor acetylhydrolase, isoform 1b, subunit 1
Gene Aliases
LIS-1, Lis1, MMS10-U, Mdsh, Ms10u, Pafaha
Chromosome Location
Chr. 11: 74673949 - 74724384 on Build GRCm38
UniGene ID
Mm.397111
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013625.4 NP_038653.1 10 1667
GenBank mRNA AK144908.1 - 10 1656
AK147207.1 - 10 1605
AK147229.1 - 9 3005
AK147237.1 - 10 1659
AK147309.1 - 10 1602
AK147507.1 - 10 1941
AK147515.1 - 10 1654
AK147533.1 - 10 1634
AK147793.1 - 10 1329
AK164008.1 - 10 1338
AK167448.1 - 10 1634
AK167824.1 - 10 1659
AK168929.1 - 10 1307
AK168960.1 - 10 1622
AY189217.1 - 9 1286
AY189218.1 - 9 1286
BC014831.1 - 10 1606 4507474
BC026141.1 - 10 1631 4017963
BC046989.1 - 10 1639
BC099462.1 - 10 1605
L25108.1 - 10 1567
L25109.1 - 6 763
U95116.1 - 10 1962

View Full Product Details