Gene Transcripts

Gene Symbol
TFRC
Entrez Gene ID
7037
Gene Name
transferrin receptor
Gene Aliases
CD71, IMD46, T9, TFR, TFR1, TR, TRFR, p90
Chromosome Location
Chr. 3: 196049284 - 196082161 on Build GRCh38
UniGene ID
Hs.529618
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128148.2 NP_001121620.1 3 193
NM_003234.3 NP_003225.2 3 334
GenBank mRNA AB209254.1 - 3 97
AK291723.1 - 3 334
AK309923.1 - 3 122
BC001188.1 - 3 107 3354176
CF994006.1 - 3 221
DA460386.1 - 3 193
M11507.1 - 3 146
X01060.1 - 3 314

View Full Product Details