Gene Transcripts

Gene Symbol
Reg2
Entrez Gene ID
19693
Gene Name
regenerating islet-derived 2
Gene Aliases
RegII
Chromosome Location
Chr. 6: 78405155 - 78408097 on Build GRCm38
UniGene ID
Mm.46360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009043.1 NP_033069.1 4 400
GenBank mRNA AK008196.1 - 4 399
BC098503.1 - 3 383 6431377

View Full Product Details