Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013667.2 | NP_038695.1 | 3, 4 | 762 | |
GenBank mRNA | AJ006036.1 | - | 3, 4 | 762 | |
AK052658.1 | - | 3, 4 | 759 | ||
AK143892.1 | - | 3, 4 | 762 | ||
BC015250.1 | - | 3, 4 | 690 | 4242038 | |
BC069911.1 | - | 3, 4 | 709 | 4974824 |