Gene Transcripts

Gene Symbol
Tnfaip6
Entrez Gene ID
21930
Gene Name
tumor necrosis factor alpha induced protein 6
Gene Aliases
TSG-6, Tnfip6, Tsg6
Chromosome Location
Chr. 2: 52038113 - 52056681 on Build GRCm38
UniGene ID
Mm.3509
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009398.2 NP_033424.1 2 236
GenBank mRNA AK139985.1 - 2 246
BC021155.1 - 2 215 4208738
U83903.1 - 2 236

View Full Product Details